Imprimer

1,3,5-Triiodo-2,4,6-trimethylbenzene 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01128178-250 BD01128178-1G
BLDPBD01128178-250EA 23 EUR
BLDPBD01128178-250 BLDPBD01128178-1G
1,3,5-Triiodo-2,4,6-trimethylbenzene 98%
1,3,5-Triiodo-2,4,6-trimethylbenzene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₉H₉I₃
Poids moléculaire: 497,8812 g/mol
Température de stockage: Ambient
Numéro CAS: 19025-36-6

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR