Imprimer

1-(3-(Difluoromethyl)-1-methyl-1H-pyrazol-4-yl)ethanone 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01155281-100
BLDPBD01155281-100EA 51 EUR
BLDPBD01155281-100
1-(3-(Difluoromethyl)-1-methyl-1H-pyrazol-4-yl)ethanone 97%
1-(3-(Difluoromethyl)-1-methyl-1H-pyrazol-4-yl)ethanone

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formule: C₇H₈F₂N₂O
Poids moléculaire: 174,1480 g/mol
Température de stockage: Freezer
Numéro CAS: 1814920-62-1

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR