Imprimer

(S)-Ethyl 2-amino-4-fluoro-4-methylpentanoate hydrochloride 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01157253-1G BD01157253-25G BD01157253-250 BD01157253-5G
BLDPBD01157253-1GEA 157 EUR
BLDPBD01157253-1G BLDPBD01157253-25G BLDPBD01157253-250 BLDPBD01157253-5G
(S)-Ethyl 2-amino-4-fluoro-4-methylpentanoate hydrochloride 97%
(S)-Ethyl 2-amino-4-fluoro-4-methylpentanoate hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formule: C₈H₁₇ClFNO₂
Poids moléculaire: 213,6775 g/mol
Température de stockage: Refrigerator
Numéro MDL: MFCD25541983
Numéro CAS: 2095926-24-0

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR