Imprimer

4-(Fluorosulphonyl)-1-methyl-1H-pyrrole-2-carboxylic acid 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01167901-100 BD01167901-250
BLDPBD01167901-100EA 92 EUR
BLDPBD01167901-100 BLDPBD01167901-250
4-(Fluorosulphonyl)-1-methyl-1H-pyrrole-2-carboxylic acid 98%
4-(Fluorosulphonyl)-1-methyl-1H-pyrrole-2-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formule: C₆H₆FNO₄S
Poids moléculaire: 207,1795 g/mol
Température de stockage: Freezer
Numéro CAS: 2138238-83-0

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR