Imprimer

(E)-1,2-Bis(4-(1H-imidazol-1-yl)phenyl)-1,2-diphenylethene 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01171081-250 BD01171081-5G BD01171081-1G
BLDPBD01171081-250EA 26 EUR
BLDPBD01171081-250 BLDPBD01171081-5G BLDPBD01171081-1G
(E)-1,2-Bis(4-(1H-imidazol-1-yl)phenyl)-1,2-diphenylethene 97%
(E)-1,2-Bis(4-(1H-imidazol-1-yl)phenyl)-1,2-diphenylethene

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₃₂H₂₄N₄
Poids moléculaire: 464,5598 g/mol
Température de stockage: Ambient
Numéro CAS: 2828431-94-1

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR