Imprimer

N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-(trifluoromethyl)benzenesulphonamide 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01176328-1G BD01176328-100 BD01176328-250 BD01176328-5G
BLDPBD01176328-1GEA 73 EUR
BLDPBD01176328-1G BLDPBD01176328-100 BLDPBD01176328-250 BLDPBD01176328-5G
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-(trifluoromethyl)benzenesulphonamide 97%
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-(trifluoromethyl)benzenesulphonamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₂₁H₁₉F₃N₂O₂S
Poids moléculaire: 420,4480 g/mol
Température de stockage: Ambient
Numéro CAS: 1105576-13-3

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR