Imprimer

Methyl methoxycarbamate 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01182202-5G BD01182202-100 BD01182202-1G BD01182202-250
BLDPBD01182202-5GEA 601 EUR
BLDPBD01182202-5G BLDPBD01182202-100 BLDPBD01182202-1G BLDPBD01182202-250
Methyl methoxycarbamate 97%
Methyl methoxycarbamate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formule: C₃H₇NO₃
Poids moléculaire: 105,0926 g/mol
Température de stockage: Refrigerator
Numéro MDL: MFCD26794522
Numéro CAS: 66508-91-6

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR