Imprimer

3-Amino-3-methyl-1-morpholinobutan-1-one hydrochloride 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01186791-100 BD01186791-250 BD01186791-1G
BLDPBD01186791-100EA 181 EUR
BLDPBD01186791-100 BLDPBD01186791-250 BLDPBD01186791-1G
3-Amino-3-methyl-1-morpholinobutan-1-one hydrochloride 97%
3-Amino-3-methyl-1-morpholinobutan-1-one hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formule: C₉H₁₉ClN₂O₂
Poids moléculaire: 222,7124 g/mol
Température de stockage: Ambient
Numéro CAS: 1415898-61-1

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR