Imprimer

5-Chloro-4-methylthiophene carboxylic acid 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01190138-250 BD01190138-100 BD01190138-1G
BLDPBD01190138-250EA 138 EUR
BLDPBD01190138-250 BLDPBD01190138-100 BLDPBD01190138-1G
5-Chloro-4-methylthiophene carboxylic acid 98%
5-Chloro-4-methylthiophene carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₆H₅ClO₂S
Poids moléculaire: 176,6207 g/mol
Température de stockage: Refrigerator
Numéro MDL: MFCD01830083
Numéro CAS: 74598-03-1

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR