Imprimer

(2R,3aR,6S,7aR)-3a-Methyl-2-((perfluorophenyl)thio)-6-(prop-1-en-2-yl)hexahydrobenzo[d][1,3,2]oxathiaphosphole 2-sulphide 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01199352-250 BD01199352-100 BD01199352-1G
BLDPBD01199352-250EA 143 EUR
BLDPBD01199352-250 BLDPBD01199352-100 BLDPBD01199352-1G
(2R,3aR,6S,7aR)-3a-Methyl-2-((perfluorophenyl)thio)-6-(prop-1-en-2-yl)hexahydrobenzo[d][1,3,2]oxathiaphosphole 2-sulphide 98%
(2R,3aR,6S,7aR)-3a-Methyl-2-((perfluorophenyl)thio)-6-(prop-1-en-2-yl)hexahydrobenzo[d][1,3,2]oxathiaphosphole 2-sulphide
Numéro CAS: 2245335-71-9

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR