Imprimer

(R)-2-(2-((R)-(4-(tert-Butyl)phenyl)sulphinyl)phenyl)-4-phenyl-4,5-dihydrooxazole 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01203465-250 BD01203465-100 BD01203465-1G
BLDPBD01203465-250EA 223 EUR
BLDPBD01203465-250 BLDPBD01203465-100 BLDPBD01203465-1G
(R)-2-(2-((R)-(4-(tert-Butyl)phenyl)sulphinyl)phenyl)-4-phenyl-4,5-dihydrooxazole 98%
(R)-2-(2-((R)-(4-(tert-Butyl)phenyl)sulphinyl)phenyl)-4-phenyl-4,5-dihydrooxazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₂₅H₂₅NO₂S
Poids moléculaire: 403,5365 g/mol
Température de stockage: Refrigerator
Numéro CAS: 2828432-14-8

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR