2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
BLDPBD01374005-25GEA
19
GBP
BLDPBD01374005-25G
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₁₅BO₃ MW: 158.01 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03093086 CAS Number: 1195-66-0 |