Benzo[b]thiophene-7-carboxaldehyde 95%

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Benzo[b]thiophene-7-carbaldehyde

BLDPBD115713-1GEA 197 GBP
BLDPBD115713-1G BLDPBD115713-250MG
Benzo[b]thiophene-7-carboxaldehyde 95%
Benzo[b]thiophene-7-carboxaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₉H₆OS
MW: 162.2083 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08572207
CAS Number: 10134-91-5

Order Now

Learn more