Benzoic acid 99%

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BLDPBD115773-5GEA 6 GBP
BLDPBD115773-5G BLDPBD115773-25G
Benzoic acid 99%
Benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₆O₂
MW: 122.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0

Order Now

Learn more