3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one 98%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
Synonyms:
1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal)
BLDPBD13444-5GEA
13
GBP
BLDPBD13444-5G
BLDPBD13444-1G
BLDPBD13444-25G
BLDPBD13444-10G
BLDPBD13444-100G
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one 98%
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₁H₁₈O₃ MW: 198.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006652 CAS Number: 69225-59-8 |