1-Boc-4-piperidineacetic acid 97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
Synonyms:
2-(1-(tert-Butoxycarbonyl)piperidin-4-yl)acetic acid
, [1-(tert-Butoxycarbonyl)piperidin-4-yl]acetic acid
BLDPBD14275-100GEA
239
GBP
BLDPBD14275-100G
BLDPBD14275-5G
BLDPBD14275-1G
BLDPBD14275-25G
BLDPBD14275-10G
BLDPBD14275-0.025G
1-Boc-4-piperidineacetic acid 97%
1-Boc-4-piperidineacetic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₁₂H₂₁NO₄ MW: 243.3000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00800239 CAS Number: 157688-46-5 |