Piperidine-4-sulphonyl chloride, N-CBZ protected 95%

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-[(Benzyloxy)carbonyl]piperidine-4-sulphonyl chlorideBenzyl 4-(chlorosulphonyl)piperidine-1-carboxylate

BLDPBD14788-100MGEA 30 GBP
BLDPBD14788-100MG BLDPBD14788-1G BLDPBD14788-250MG BLDPBD14788-5G BLDPBD14788-10G
Piperidine-4-sulphonyl chloride, N-CBZ protected 95%
Piperidine-4-sulphonyl chloride, N-CBZ protected

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₃H₁₆ClNO₄S
MW: 317.788 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05664659
CAS Number: 287953-54-2

Order Now

Learn more