Imprimer

Folinic acid calcium salt hydrate (Leucovorin calcium) 95%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BLDPBD18473-5GEA 153 EUR
BLDPBD18473-5G BLDPBD18473-1G BLDPBD18473-250MG
Folinic acid calcium salt hydrate (Leucovorin calcium) 95%
Folinic acid calcium salt hydrate (Leucovorin calcium)

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₂₀H₂₁CaN₇O₇·xH₂O
Poids moléculaire: 511,51 g/mol (anhy)
Température de stockage: Température ambiante
Numéro MDL: MFCD08704249
Numéro CAS: 1492-18-8

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR