1,3-Di-4-piperidylpropane 98%

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,4'-Trimethylenedipiperidine1,3-Bis(4-piperidyl)propane

BLDPBD18807-1GEA 6 GBP
BLDPBD18807-1G
1,3-Di-4-piperidylpropane 98%
1,3-Di-4-piperidylpropane
Formula: C₁₃H₂₆N₂
MW: 210.37 g/mol
MDL Number: MFCD00038013
CAS Number: 16898-52-5

Order Now

Learn more