3'-Phenoxyacetophenone 99%

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(3-Phenoxyphenyl)ethan-1-one1-(3-Phenoxyphenyl)ethanone

BLDPBD223918-5GEA 160 GBP
BLDPBD223918-5G BLDPBD223918-1G BLDPBD223918-25G
3'-Phenoxyacetophenone 99%
3'-Phenoxyacetophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₄H₁₂O₂
MW: 212.2439 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08543460
CAS Number: 32852-92-9

Order Now

Learn more