5-Fluoro-2-(methylcarbamoyl)phenylboronic acid 98%

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (5-Fluoro-2-(methylcarbamoyl)phenyl)boronic acid

BLDPBD230984-250MGEA 89 GBP
BLDPBD230984-250MG BLDPBD230984-1G BLDPBD230984-100MG
5-Fluoro-2-(methylcarbamoyl)phenylboronic acid 98%
5-Fluoro-2-(methylcarbamoyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₉BFNO₃
MW: 196.9700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878343
CAS Number: 957062-96-3

Order Now

Learn more