10-Undecynoic acid 98%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
Synonyms:
Undec-10-ynoic acid
BLDPBD33010-100MGEA
24
GBP
BLDPBD33010-100MG
BLDPBD33010-250MG
BLDPBD33010-5G
BLDPBD33010-25G
BLDPBD33010-1G
10-Undecynoic acid 98%
10-Undecynoic acid
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₁H₁₈O₂ MW: 182.26 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00014389 CAS Number: 2777-65-3 |