Imprimer

L-Lysine 97% + (contains < 5% H₂O)

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): L(+)-Lysine(S)-(+)-LysineH-Lys-OH

BLDPBD34122-1000GEA 41 EUR
BLDPBD34122-1000G BLDPBD34122-500G BLDPBD34122-25G BLDPBD34122-10G BLDPBD34122-100G BLDPBD34122-5G
L-Lysine 97% + (contains < 5% H₂O)
L-Lysine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₁₄N₂O₂
Poids moléculaire: 146,19 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00064433
Numéro CAS: 56-87-1

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR