m-PEG11-amine 98+%

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5,8,11,14,17,20,23,26,29,32-Undecaoxatetratriacontan-34-amine

BLDPBD434644-250MGEA 167 GBP
BLDPBD434644-250MG BLDPBD434644-100MG BLDPBD434644-1G
m-PEG11-amine 98+%
m-PEG11-amine

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₃H₄₉NO₁₁
MW: 515.635 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27977521
CAS Number: 854601-60-8

Order Now

Learn more