4-Bromo-2-fluorobenzylamine hydrochloride 98%

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Bromo-2-fluorophenyl)methanamine hydrochloride

BLDPBD4734-5GEA 62 GBP
BLDPBD4734-5G BLDPBD4734-1G BLDPBD4734-25G
4-Bromo-2-fluorobenzylamine hydrochloride 98%
4-Bromo-2-fluorobenzylamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₇BrFN·HCl
MW: 240.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143266
CAS Number: 147181-08-6

Order Now

Learn more