Methyl-5-chlorosalicylate 98%

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 5-chloro-2-hydroxybenzoate

BLDPBD6116-500GEA 166 GBP
BLDPBD6116-500G BLDPBD6116-5G BLDPBD6116-10G BLDPBD6116-100G BLDPBD6116-1000G BLDPBD6116-25G
Methyl-5-chlorosalicylate 98%
Methyl 5-chlorosalicylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₇ClO₃
MW: 186.60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016461
CAS Number: 4068-78-4

Order Now

Learn more