4-(Dimethylcarbamoyl)phenylboronic acid pinacol ester 98%

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(N,N-Dimethylaminocarbonyl)phenylboronic acid, pinacol ester4-(Dimethylcarbamoyl)phenylboronic Acid Pinacol Ester

BLDPBD8066-1GEA 26 GBP
BLDPBD8066-1G BLDPBD8066-250MG BLDPBD8066-5G
4-(Dimethylcarbamoyl)phenylboronic acid pinacol ester 98%
4-(Dimethylcarbamoyl)phenylboronic acid pinacol ester

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₅H₂₂BNO₃
MW: 275.1511 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863908
CAS Number: 400727-57-3

Order Now

Learn more