Imprimer
Papier de protection de surface, Benchkote® Plus, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Whatman™ Benchkote Plus™
115-9210EA 291 EUR
115-9210 115-9211
Papier de protection de surface, Benchkote® Plus, Whatman™
Protection des surfaces Protections pour paillasse
Cytiva's Whatman™ Benchkote® Plus bench protector is a thicker, more absorbent version of the original Benchkote® bench paper. Designed for demanding or hazardous applications, it can absorb more than 750 ml/m² water.
  • Material is strong and tear resistant when wet or dry
  • Smooth white surface can be written on with ink or pencil and lies flat
  • Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
  • Can be incinerated after use: PE layer is rapidly consumed when incinerated and does not melt or drip
Use of the robust Benchkote® Plus bench liner can help prevent many common sources of laboratory contamination, such as those involving hazardous materials on shared workspace benches. It can also help with disposal of hazardous materials.