Imprimer
Votre recherche pour: Fioles
309  résultat(s)
SearchResultCount:"309"
Sort Results
Vue liste Nouvelle Vue
Evaluer les résultats de votre recherche
Fournisseur: VWR Collection
Description: Short thread ND9 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropyle...
Fournisseur: VWR Collection
Description: Designed for freezing samples in all cryopreservation applications, these vials feature an internal thread with washer, or an external thread.
promo
Fournisseur: VWR Collection
Description: VWR®, Vial insert, 0,10 ml, Verre, incolore, Ø×h: 6×31 mm, Fond conique, Non stérile, Cap included: Non
Fournisseur: VWR Collection
Description: ND11 crimp neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of aluminiu...
Fournisseur: VWR Collection
Description: Headspace ND18 vials with screw neck manufactured from 1st hydrolytic class borosilicate glass with a round bottom. They can be purchased with a varie...
Fournisseur: VWR Collection
Description: Snap ring ND11 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polyethylene ...
Fournisseur: VWR Collection
Description: ND8 screw neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropy...
Fournisseur: VWR Collection
Description: ND24 screw neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polyprop...
Fournisseur: VWR Collection
Description: ND13 screw neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polyprop...
Fournisseur: VWR Collection
Description: VWR®, Vial insert, 0,10 ml, Verre, incolore, Ø×h: 5×31 mm, Fond conique, Non stérile, Cap included: Non
Fournisseur: VWR Collection
Description: Shell vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a PE plug featuring a starburst top...
Fournisseur: VWR Collection
Description: Headspace ND20 vials with crimp neck manufactured from 1st hydrolytic class borosilicate glass in both flat bottom and round bottom style. They can be...
Fournisseur: VWR Collection
Description: Pour le stockage, la manipulation et le transport des mouches drosophiles.
Référence Produit: (548-0859A)
Fournisseur: VWR Collection
Description: Crimp Neck ND13 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of aluminium se...
UOM: 1 * 1.000 Pce
Fournisseur: VWR Collection
Description: En verre sodo-calcique, ambré.
Fournisseur: VWR Collection
Description: Ces flacons sont utilisés de préférence sur des instruments des fabricants suivants: Agilent, Beckman, Carlo Erba, CTC, Fisons, PerkinElmer, Shimadzu,...
Prix sur demande
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 01 45 14 89 12.
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 01 45 14 89 12.
Ces articles ne peuvent pas être ajoutés à votre panier en raison des exigences réglementaires. Veuillez envoyer un e-mail à webshop.fr@vwr.com
Veuillez noter qu'une autorisation peut être nécessaire pour commander ce produit. Un représentant de VWR vous contactera si votre commande le nécessite.
Ce produit a été bloqué par votre entreprise. Veuillez s'il vous plait contacter votre service achats pour plus d'informations.
Le produit original n'est plus disponible. Le remplacement représenté est disponible
Ce produit n'est plus disponible. Des alternatives peuvent être disponibles en faisant une recherche avec la référence VWR ci-dessus. Si vous avez besoin d'aide, s'il vous plaît appelez le Service Clients au 0825 02 30 30 (0,18 € TTC/mn).
1 - 16 of 309
no targeter for Bottom