Imprimer
VWR®, Verres de montre
Fournisseur: VWR
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VWR®
216-0161EA 26.4 EUR
216-0161 216-0162 216-0160 216-0155 216-0163 216-0455 216-0158 216-0159 216-0156 216-0454 216-0157
VWR®, Verres de montre
Verres de montre
PTFE.
  • Excellent chemical resistance
  • Temperature resistant from –200 to 280 °C
  • Exceptionally smooth surfaces to prevent contamination
Ideal for covering PTFE beakers when decomposing and evaporating, can also be used as evaporating dishes. Extremely robust.