Digital polarimeter, Polax-2L
Supplier: Atago
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
635-0162EA 0 GBP
ATAG5223
Digital polarimeter, Polax-2L
Digital polarimeter for use in the pharmaceutical, chemical and food industries.
  • Long lasting LED light source with interference filter
  • Dual scale either angle of rotation or International sugar scale can be selected
  • Temperature of sample chamber can be displayed
The robust and simple design is suited to educational applications for students to aid understanding of optical rotation of polarised light.
Delivery information: Supplied with 100 mm and 200 mm observation tubes.