Imprimer
Thermomètre numérique de précision, testo 110
Fournisseur: Testo
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
620-1198EA 152 EUR
620-1198
Thermomètre numérique de précision, testo 110
Thermomètres Thermomètres numériques
Thanks to the NTC technology tucked away inside this thermometer, it can measure temperatures in very low temperature environments.
  • Extremely accurate, also in very low temperature ranges, i.e. cold storage rooms
  • Optional connectable temperature sensor and wireless sensor for different applications in the field
  • User-defined thresholds; audible alarm alerts when thresholds are exceeded
  • Large, two-line display
  • Min/max value recognition
  • Auto-hold, freezes the last reading
Ideal for use in sold storage rooms, cold storage warehouses and outdoors.
Certifications: HACCP compliant, certified according to EN 13485 for use in the food sector.
Informations de livraison: Fourni avec pile 6F22 de neuf volts et protocole d'étalonnage, sans sonde. Les sondes peuvent être commandées séparément.