Imprimer
Pressure vessels
Fournisseur: Cytiva (Formerly Pall Lab)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
516-9051EA 9060 EUR
PALL15203
Pressure vessels
The materials used for construction are corrosion resistant, with broad chemical compatibility. Multiple ports allow for temperature probes, pressure gauges or other monitoring devices.
  • Non-sterile and long vessel life
  • Maximum operating pressure: 7 bar
  • Wide-mouth opening makes clean-up easy
These vessels are ideal for processing of batches of liquids for sterilisation or clarification. These vessels designed in a manner to resist baking and electropolished finish provides ultra-smooth walls.
Informations de livraison: Includes with pressure gauge, safety relief valve, flow shut-off valve and hose barb connectors.