Imprimer
Flacons, col étroit
Fournisseur: Kartell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
KART1684EA 129 EUR
KART1684 KART1683 KART1686 KART1687
Flacons, col étroit
PFA bottle, ETFE screw cap.
  • Temperature resistance: −200 to +250 °c
  • Very good chemical resistance
A special sealing lip on the cap has been developed for these bottles to ensure a leakproof closure.