Imprimer
Unités de distillation Kjeldahl, série UDK
Fournisseur: VELP Scientifica
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
701-0227EA 0 EUR
VELPF30200150
Unités de distillation Kjeldahl, série UDK
The UDK 159 combines all the advantages of a fully automatic distillation with the added benefits of integrated colorimetric titration (AOAC approved). It runs automatically, after setting sodium hydroxide, water and boric acid addition, the steam generation output from 10 to 100%. The high precision pumps and burette ensure constant accurate dosing of reagents and with the integrated colorimetric titrator (AOAC recommended) you will have reliable results concerning your nitrogen and protein determinations.
  • Automatic NaOH, H₂O and H₃BO₃ addition
  • Delay time (Devarda alloy analysis)
  • Washing and blank analysis
  • Automatic titration vessel washing
  • Distillation and titration residues removal; distillation in series
  • Balance connection
  • Language selection and electronic user guide
  • Safety lever and sensors and reagent level warning to protect the user
All UDK series distillation units have an alkali resistant techno polymer housing and accept different kinds of test tubes: straight tubes (100, 250, 400 ml and 1 L) or Kjeldahl balloon (500 ml).
Certifications: Conformity with multiple standards protocols AOAC, ISO, EPA, DIN etc.
Informations sur les accessoires: Optional accessories such as test tubes, test tube spacer, test tube connection, connection cables and IQ/OQ/PQ manuals are available on request.
Informations de livraison: Each unit comes ready to use and is supplied with 250 ml test tube, 250 ml collecting flask, pincer for test tubes, touch pen and protective film for touch screen.