Imprimer
Reaction vessels, round bottom, with flat flange, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BOLA
BOHLB281-03EA 881 EUR
BOHLB281-03 BOHLB281-15 BOHLB281-12 BOHLB281-06 BOHLB281-09
Reaction vessels, round bottom, with flat flange, BOLA
Récipients de réaction Flacons à réactions
PTFE, white.
  • Temperature resistance: from −200 to +200 °C
  • Universal chemical resistance
  • Vacuum: suitable
Round bottom, thick wall, smooth interior surface. Can be heated by a thermostat or an electric heating mantle.