Imprimer
Flat flange reaction vessel, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BOLA
BOHLB320-01EA 1790 EUR
BOHLB320-01
Flat flange reaction vessel, BOLA
Récipients de réaction Béchers à rodage plan
PFA, translucent.
  • Temperature resistance from −200 to +250°C
  • Very good chemical resistance
  • FDA conform
Flat bottom, non-porous, big opening. Can be heated by thermostats or an electric heating mantle or can be used with a heatable magnetic stirrer and a PTFE-encapsulated magnetic stirring bar for stirring.