Imprimer
Screw-in tubing connectors, straight, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BOLA
BOHLD579-02EA 65.6 EUR
BOHLD579-02 BOHLD579-24 BOHLD579-22 BOHLD579-06 BOHLD579-26 BOHLD579-04
Screw-in tubing connectors, straight, BOLA
Raccords et connecteurs
Straight fitting, made of PTFE, with one connector for elastic tubing (e.g. Viton®, Tygon®, silicone) and one screw-in thread (either NPT or G).
  • Temperature resistance from −200 to +250 °C
  • Universal chemical resistance
  • Suitable for vacuum