Imprimer
Banded earplugs, QB1HYG®
Fournisseur: Honeywell Safety
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DALL3301282EA 85.92 EUR
DALL3301282
Banded earplugs, QB1HYG®
These foam earplugs offer solid protection with supra-aural fit for people who are exposed to intermittent noise.
  • Smooth, ergonomic pods fit inside the opening of the ear canal for maximum protection
  • Bright colours for improved visibility
  • Lightweight and portable
SNR: 27 dB
Certifications: EN 352-2
Cdt: In a resealable bag.