Imprimer
Examination gloves, Profile 3000™
Fournisseur: Ansell-Nitritex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Profile™
NITRPNA3103-MEA 505 EUR
NITRPNA3103-M NITRPNA3104-L NITRPNA3101-XS NITRPNA3106-XXL NITRPNA3102-S NITRPNA3105-XL NITRPNA3107-XXXL
Examination gloves, Profile 3000™
Gants
These powder-free ambidextrous nitrile gloves provide a good resistance against a wide range of chemical as well as against punctures and are ideal for use in medical environments.
  • Latex-free to eliminate Type 1 allergies
  • Beaded cuff provides easier donning and stability on the arm
  • Textured finish for improved grip
  • Latex-free to eliminate Type 1 allergies
  • Beaded cuff provides easier donning and stability on the arm
  • Textured finish for improved grip
  • Packed in practical dispenser box
Certifications: Complies with the requirements of European Glove Standards EN420:2003 +A1:2009 and EN374-1:2016, EN374-5:2016 and EN421:2010 Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications, suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006. Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Cdt: Each size carries a unique bar code on the inner dispenser box (EAN-13) and the carton (ITF-14) to allow automatic data entry into Stock Control Systems.