Imprimer
Filtres
Fournisseur: Olympus
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
630-0958EA 0 EUR
630-0958 630-0947 630-0948 630-0959 630-0957 630-0946 630-0961 630-0960 630-0949
Filtres
Filtres Filtres optiques
Filter units, ranging from UV excitation to RFP, respond to applications using various fluorescent dyes and protein.
High quality (HQ) filters have an edge steepness and high transmission that efficiently detect the light of fluorescence to enhance and capture brighter fluorescence images in precise detail.