Screw thread vials, Target®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Target®
NSCAC4010-LV1EA 166 GBP
NSCAC4010-LV1 NSCAC4010-2 NSCAC4010-V1
Screw thread vials, Target®
Vials
Vials feature a 40% larger opening than standard vials and a uniformly flat bottom for security with inserts. Made from Type I, Class A borosilicate glass. Target I-D™ vials have a ceramic write-on ID patch with graduations at 0,5, 1,0 and 1,5 ml. Silanised vials help to limit protein adsorption.
Inserts are designed for wide opening 2 ml vials. Flat bottom MicroSert™ inserts are made from borosilicate glass. Conical-style Polyspring® inserts are available in borosilicate glass or PP.