Imprimer
Organisateur de tiroir, Lab Fridge™
Fournisseur: Bel-Art Products
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Scienceware®
212-6601EA 217 EUR
212-6601 212-6603 212-6602
Organisateur de tiroir, Lab Fridge™
Plateaux/Récipients/Pots Plateaux à tiroirs
Cabinet made of PP and holds PS racks.
  • Saves space
  • Eliminates clutter and allows easy stock rotation
  • Accommodates bottles with a maximum height of 83 mm (3¹/₄")
  • Can be used in freezers with temperatures as low as −40 °C
Suitable for refrigerated storage of small bottles of blood-typing sera, chemistry controls, enzyme reagents, and coagulant reagents.
Informations de commande: Cabinets and racks can be ordered separately.