Imprimer
Tubing couplings, rapid, NW 6,4 mm
Fournisseur: Bürkle
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BURK8761-1050EA 19.9 EUR
BURK8761-1050 BURK8761-2160 BURK8771-1140 BURK8761-1150 BURK8761-2140 BURK8771-1040 BURK8775-2140 BURK8771-1160 BURK8771-1060 BURK8760-1141 BURK8763-1040 BURK8760-1142 BURK8763-1140 BURK8760-1061 223-1133 BURK8773-1150 BURK8770-2142 BURK8763-2160 BURK8772-1380 BURK8761-1160 BURK8761-1060 BURK8761-1140 BURK8761-1040 BURK8771-1150 BURK8771-2140 BURK8775-1160 BURK8771-1050 BURK8775-1040 BURK8771-2160 BURK8760-2141 BURK8763-1150 BURK8760-2161 BURK8773-1160 BURK8773-1140 BURK8770-1042 BURK8772-1310 BURK8770-1142 BURK8762-1380 BURK8763-1160 BURK8762-1140 BURK8770-1041
Tubing couplings, rapid, NW 6,4 mm
Raccords et connecteurs
Spring/lock Stainless steel V4A
Fast and safe connection for liquids, gases, compressed air and vacuum. Rapid couplings are the ideal when connecting and separating hoses, machines, apparatus, containers, etc. Top quality, with tested reliability, small space requirement, maximum possible flow, easy operation (single handed operation) and an elegant design distinguish the rapid couplings.The couplings are available in two different materials: POM (Polyoxymethylene) and PP (Polypropylene). The rapid couplings are available with a hose nozzle, hose screw joint, thread connector or as a built-in component for control panels, with or without stop-valve respectively.The couplings are simply connected to each other, automatically clicking intoplace and opening the valves.When disconnecting, (one handed operation), the valves are automatically sealed, no leakage occurs.
POM
Seal (standard): Buna-N
Operating pressure at 20 °C with vacuum up to 10 bar
Operating temperature: −40 to +80 °C
PP
Seal (standard): EPDM
Operating pressure at 20 °C: up to 8 bar
Operating temperature: 0 to +110 °C