Imprimer
Tampon SDS TRIS-glycine, poudre, proteomics grade
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VWR®
M149-40LEA 122 EUR
VWRCM149-40L
Tampon SDS TRIS-glycine, poudre, proteomics grade
The buffers used for protein electrophoresis maintain the pH and temperature of the environment as well as provide ions that carry electrical current. There are a variety of buffers available, although Tris-Glycine (TG) and TG-SDS have been the standards since their introduction.
Chaque conditionnement permet de préparer 1 l de concentré 10X. Le conditionnement de 40 l contient suffisamment de poudre sèche pour préparer 40 litres de tampon 1X. La solution 1X contient : 0,025 M de tris, 0,192 M de glycine et 0,1% de SDS.