Imprimer
TRIS-tricine Solution tampon 10x concentrée, grade dédié à la protéomique
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M170-1LEA 241 EUR
VWRCM170-1L
TRIS-tricine Solution tampon 10x concentrée, grade dédié à la protéomique
The Tris-Tricine (TT) buffer system is a popular replacement for Tris-Glycine buffers because it stacks and resolves small polypeptides with greater efficiency than TG buffers.
  • Convenient concentrations and packaging for common protein gel electrophoresis buffers
  • Excellent quality, consistency and resolution