Imprimer
Trousers, Fristads® PR54-220, Design A, navy blue
Fournisseur: Fristads Kansas Group
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Fristads®
113-00692EA 0 EUR
113-00692 113-00691 113-00694 113-00693 113-00690 113-00612 113-00611 113-00614 113-00613 113-00695 113-00610 113-00609 113-00608 113-00605 113-00604 113-00607 113-00606 113-00623 113-00622 113-00589 113-00625 113-00624 113-00586 113-00588 113-00621 113-00620 113-00587 113-00619 113-00616 113-00615 113-00618 113-00617 113-00593 113-00592 113-00595 113-00594 113-00591 113-00590 113-00634 113-00633 113-00635 113-00630 113-00597 113-00596 113-00599 113-00632 113-00631 113-00598 113-00627 113-00626 113-00628 113-00647 113-00646 113-00650 113-00656 113-00655 113-00658 113-00657 113-00652 113-00651 113-00654 113-00653 113-00648 113-00661 113-00660 113-00667 113-00666 113-00669 113-00668 113-00663 113-00662 113-00665 113-00664 113-00659 113-00670 113-00672 113-00671 113-02570 113-00678 113-00677 113-00679 113-00674 113-00673 113-00676 113-00675 113-00681 113-00680 113-00683 113-00682 113-00601 113-00689 113-00688 113-00600 113-00603 113-00602 113-00685 113-00684 113-00687 113-00686
Trousers, Fristads® PR54-220, Design A, navy blue
Tenue de travail
Thanks to their durability and comfort, these trousers are perfectly suitable for use in garages as well as in industrial and service companies. The material is made of 65% polyester and 35% cotton. The inside is brushed and provides a comfortable cotton feel. The outside is very hardwearing and does not bleach, even after countless washes.
  • Two front and back pockets
  • Two leg pockets - one with flap, telephone pocket, pen pocket, tool bags
  • Adjustable waist, plastic snap buttons
  • Colour accents on pocket edges
Weight: 260 g/m²
Basic colour: navy blue