Imprimer
Trousers, Fristads® PR54-220, Design A, dark grey
Fournisseur: Fristads Kansas Group
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Fristads®
113-00854EA 0 EUR
113-00854 113-00853 113-00856 113-00855 113-00850 113-00852 113-00851 113-00847 113-00846 113-00849 113-00848 113-00865 113-00864 113-02649 113-00867 113-00900 113-00866 113-00861 113-00860 113-00863 113-00862 113-00858 113-00857 113-00859 113-02651 113-02652 113-00870 113-02650 113-00876 113-02659 113-00875 113-00911 113-00878 113-02657 113-00877 113-00910 113-02658 113-02655 113-00872 113-02656 113-00871 113-00874 113-02653 113-00873 113-02654 113-00906 113-00905 113-00908 113-00907 113-00902 113-00869 113-00901 113-00868 113-00904 113-00903 113-00909 113-02662 113-02663 113-02660 113-00881 113-00880 113-02661 113-00887 113-02703 113-00920 113-02704 113-00886 113-02668 113-00922 113-02701 113-00889 113-00921 113-02669 113-02702 113-00888 113-00883 113-02666 113-02700 113-02667 113-00882 113-00885 113-02664 113-02665 113-00884 113-00917 113-00916 113-00919 113-00918 113-00913 113-00879 113-00912 113-00915 113-00914 113-00890 113-02673 113-02674 113-02671 113-00892 113-02672 113-00891 113-02670 113-00931 113-02714 113-00898 113-02715 113-00897 113-00930 113-02679 113-00933 113-02712 113-00932 113-02713 113-00899 113-02677 113-00894 113-02710 113-00893 113-02678 113-02711 113-00896 113-02675 113-00895 113-02676 113-00928 113-00927 113-02709 113-00929 113-02707 113-00924 113-00923 113-02708 113-00926 113-02705 113-02706 113-00925 113-02684 113-02685 113-02682 113-02683 113-02680 113-02681 113-02688 113-02689 113-02686 113-02687 113-00935 113-00934 113-02716 113-02695 113-02696 113-02693 113-02694 113-02691 113-02692 113-02690 113-00832 113-00831 113-00834 113-00833 113-02699 113-02697 113-00830 113-02698 113-00829 113-00828 113-00827 113-00826 113-00843 113-00842 113-00845 113-00844 113-00841 113-00840 113-00839 113-00836 113-00835 113-00838 113-00837
Trousers, Fristads® PR54-220, Design A, dark grey
Tenue de travail
Thanks to their durability and comfort, these trousers are perfectly suitable for use in garages as well as in industrial and service companies. The material is made of 65% polyester and 35% cotton. The inside is brushed and provides a comfortable cotton feel. The outside is very hardwearing and does not bleach, even after countless washes.
  • Two front and back pockets
  • Two leg pockets - one with flap, telephone pocket, pen pocket, tool bags
  • Adjustable waist, plastic snap buttons
  • Colour accents on pocket edges
Weight: 260 g/m²
Basic colour: dark grey