Imprimer
Trousers, Fristads® PR54-220, Design A, black
Fournisseur: Fristads Kansas Group
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Fristads®
113-02750EA 0 EUR
113-02750 113-01023 113-02751 113-01024 113-01021 113-01022 113-01020 113-00975 113-00974 113-02759 113-01029 113-00977 113-00976 113-01027 113-00971 113-00970 113-01028 113-01025 113-00973 113-02752 113-01026 113-00972 113-00968 113-00967 113-02749 113-00969 113-02761 113-01034 113-01035 113-02762 113-00980 113-01032 113-01033 113-02760 113-01030 113-01031 113-00986 113-02769 113-00985 113-02767 113-00988 113-00987 113-02768 113-00982 113-01038 113-02765 113-01039 113-00981 113-02766 113-01036 113-02763 113-00984 113-02764 113-00983 113-01037 113-00979 113-00978 113-01045 113-02772 113-01046 113-02773 113-01043 113-00991 113-02770 113-01044 113-02771 113-00990 113-01041 113-01042 113-01040 113-02778 113-02779 113-00993 113-01049 113-02776 113-00992 113-02777 113-00995 113-02774 113-01047 113-02775 113-00994 113-01048 113-00989 113-01056 113-02783 113-01057 113-02784 113-01054 113-02781 113-02782 113-01055 113-01052 113-02780 113-01053 113-01050 113-01051 113-02789 113-02787 113-02788 113-02785 113-01058 113-02786 113-01059 113-02794 113-01065 113-02792 113-02793 113-01063 113-02790 113-01064 113-02791 113-01061 113-01062 113-01060 113-02725 113-02726 113-02723 113-02724 113-00953 113-02736 113-01009 113-00952 113-02737 113-00955 113-01007 113-02734 113-01008 113-02735 113-00954 113-02732 113-02733 113-01006 113-02730 113-00951 113-02731 113-00950 113-00949 113-02729 113-00946 113-02727 113-00948 113-02728 113-00947 113-01012 113-01013 113-02740 113-01010 113-01011 113-00964 113-02747 113-00963 113-02748 113-02745 113-01018 113-00966 113-01019 113-02746 113-00965 113-02743 113-01016 113-00960 113-01017 113-02744 113-00962 113-02741 113-01014 113-01015 113-02742 113-00961 113-00957 113-00956 113-00959 113-02738 113-02739 113-00958
Trousers, Fristads® PR54-220, Design A, black
Tenue de travail
Thanks to their durability and comfort, these trousers are perfectly suitable for use in garages as well as in industrial and service companies. The material is made of 65% polyester and 35% cotton. The inside is brushed and provides a comfortable cotton feel. The outside is very hardwearing and does not bleach, even after countless washes.
  • Two front and back pockets
  • Two leg pockets - one with flap, telephone pocket, pen pocket, tool bags
  • Adjustable waist, plastic snap buttons
  • Colour accents on pocket edges
Weight: 260 g/m²
Basic colour: black