Imprimer
Trousers, Fristads® PR54-220, Design B, Royal Blue
Fournisseur: Fristads Kansas Group
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Fristads®
113-01265EA 0 EUR
113-01265 113-02871 113-02872 113-01266 113-01263 113-02870 113-01264 113-01261 113-01262 113-01260 113-02879 113-02912 113-02913 113-02910 113-01304 113-02877 113-02878 113-02911 113-01269 113-01302 113-02875 113-02876 113-01303 113-01300 113-01267 113-02873 113-01301 113-02874 113-01268 113-02909 113-02907 113-02908 113-02905 113-02906 113-02903 113-02904 113-02882 113-01276 113-01277 113-02883 113-02880 113-01274 113-02881 113-01275 113-01272 113-01273 113-01270 113-01271 113-02923 113-02924 113-02888 113-02921 113-02889 113-02922 113-02886 113-02887 113-02920 113-02884 113-01278 113-01279 113-02885 113-02918 113-02919 113-02916 113-02917 113-02914 113-02915 113-02893 113-01287 113-01200 113-01288 113-02894 113-02891 113-01285 113-02892 113-01286 113-01283 113-01284 113-02890 113-01281 113-01282 113-02934 113-01207 113-01208 113-02935 113-02899 113-01205 113-02932 113-01206 113-02933 113-02930 113-01203 113-02897 113-02931 113-02898 113-01204 113-01201 113-01289 113-02895 113-02896 113-01202 113-02929 113-02927 113-02928 113-02925 113-02926 113-01280 113-01210 113-01298 113-01211 113-01299 113-01296 113-01297 113-01294 113-01295 113-01292 113-01293 113-01218 113-01219 113-01216 113-01217 113-01214 113-01215 113-01212 113-01213 113-02938 113-02936 113-01209 113-02937 113-01290 113-01291 113-01188 113-01221 113-01189 113-01222 113-01186 113-01220 113-01187 113-01185 113-01229 113-01227 113-01228 113-01225 113-01226 113-01223 113-01224 113-01232 113-01199 113-01233 113-01197 113-01230 113-01231 113-01198 113-01195 113-01196 113-01193 113-01194 113-01238 113-01239 113-01236 113-01237 113-01234 113-01235 113-01191 113-01192 113-01190 113-01243 113-01244 113-01241 113-01242 113-01240 113-01249 113-01247 113-01248 113-01245 113-01246 113-01254 113-01255 113-01252 113-01253 113-01250 113-01251 113-02901 113-02868 113-02902 113-02869 113-02900 113-02867 113-01258 113-01259 113-01256 113-01257
Trousers, Fristads® PR54-220, Design B, Royal Blue
Tenue de travail
Thanks to their durability and comfort, these trousers are perfectly suitable for use in garages as well as in industrial and service companies. The material is made of 65% polyester and 35% cotton. The inside is brushed and provides a comfortable cotton feel. The outside is very hardwearing and does not bleach, even after countless washes.
  • Two front and back pockets
  • Two leg pockets - one with flap, telephone pocket, pen pocket, tool bags
  • Adjustable waist, plastic snap buttons
  • Colour accents on pocket edges and leg pocket flap
Weight: 260 g/m²
Basic colour: royal blue